Finnpipette™ F2 Good Laboratory Pipetting (GLP) Kits
Thermo Scientific™

Finnpipette™ F2 Good Laboratory Pipetting (GLP) Kits

Enjoy the convenience of having a variety of Finnpipette™ F2 single channels packaged together, in a range of volumes, with compatible Finntip pipette tips and a pipette stand to support good laboratory pipetting in your lab
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
47008800.2 to 1000 μL
47008701 to 1000 μL
470088510 to 10,000 μL
47010702 to 1000 μL
Catalog number 4700880
Price (EUR)
909,90
Online exclusive
1.011,00
Save 101,10 (10%)
Each
Volume (Metric):
0.2 to 1000 μL
Price (EUR)
909,90
Online exclusive
1.011,00
Save 101,10 (10%)
Each

Access everything needed for pipetting in these all-inclusive kits. Thermo Scientific™ Finnpipette™ F2 GLP (Good Laboratory Pipetting) Kits provide durable, high-performance Finnpipette F2 pipettes which offer maximum reliability and comfort. Built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light, these variable volume pipettes are fully autoclavable without disassembling.

For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.

Finnpipette F2: Proven reliable for long-term intensive use

Features:

  • Fully autoclavable for protection
  • Tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light
  • Easy maintenance
  • Color coded together with Finntips for easier recognition of compatible tips

Kit 1 includes:

  • Three Thermo Scientific Finnpipette F2 Pipettes from 1 to 1000 μL:
    • 1-10 μL micro
    • 10-100 μL
    • 100-1000 μL
  • Flex tips: 10 micro: 1 x 96; 200: 1 x 96; 1000: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 2 includes:

  • Four Thermo Scientific Finnpipette F2 Pipettes from 0.2 to 1000 μL:
    • 0.2-2 μL
    • 2-20 μL univ
    • 20-200 μL
    • 100-1000 μL
  • Flex tips: 10 micro: 1 x 96; 200: 2 x 96; 1000: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 3 includes:

  • Three Thermo Scientific Finnpipette F2 Pipettes from 10 to 10,000 μL:
    • 10-100 μL
    • 100-1000 μL
    • 1000-10,000 μL
  • Flex tips: 200: 1 x 96; 1000: 1 x 96; FT 10 mL: 1 x 24
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 4 includes:

  • Three Thermo Scientific Finnpipette F2 Pipettes from 20 to 1000 μL:
    • 2-20 μL
    • 20-200 μL
    • 100-1000 μL 
  • Flex tips: 200: 2 x 96; 1000: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Ordering Information:

The kit contains samples of Finntip Flex, the most advanced traditional tip offering very low attachment and ejection forces. The Good Laboratory Pipetting Guide contains helpful tips for pipetting including pipetting techniques, ergonomic considerations and how to select tips for best performance.

Warranty: 5 years with web registration.

Specifications
Compatible TipsFlex Tips: 10, 200, 1000 μL
DescriptionFinnpipette F2 GLP Kit 2: 0.2-2μL micro, 2-20μL, 20-200μL and 100-1000μL
ModelF2
AutoclavableAutoclavable
ColorAssorted
Product LineFinnpipette F2
TypeGLP Kit
Volume (Metric)0.2 to 1000 μL
Unit SizeEach